Merck
  • Alternative eluent composition for LC-MS analysis of perfluoroalkyl acids in raw fish samples.
Alternative eluent composition for LC-MS analysis of perfluoroalkyl acids in raw fish samples.
Journal of agricultural and food chemistry (2014-05-23)
Tõiv Haljasorg, Jaan Saame, Karin Kipper, Anu Teearu, Koit Herodes, Mari Reinik, Ivo Leito
ABSTRACT
A wide range of anthropogenic pollutants that possess serious environmental and health risks are known. One type of these harmful substances that have become a focus of interest during the past decade are perfluoroalkyl acids (PFAAs), which are extensively used in industry for different purposes. Due to the harmful effects that these compounds might cause in living organisms, EFSA and EU CONTAM panel have issued a monitoring program for PFAAs in foodstuffs. This has given rise to intense research dedicated to the analysis of PFAAs over the past few years. This work focuses on chromatographic analysis of three PFAAs in fish. The analytes, perfluorooctanoic acid (PFOA), perfluorononanoic acid (PFNA), and perfluorooctanesulfonic acid (PFOS), are commonly associated with the production of fluoropolymers. Fluorinated alcohols are used as eluent components, and their possible advantages as eluent modifiers in LC-MS analysis of PFAAs, alternative retention mechanism and enhanced ionization efficiency, are examined. The analyzed fish samples originating from Estonian fresh and marine waters had low contents of PFAAs.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Supelco
1,1,1,3,3,3-Hexafluoro-2-propanol, for GC derivatization, LiChropur, ≥99.8%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
1,1,1,3,3,3-Hexafluoro-2-propanol, ≥99%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)