Merck
  • Aminopeptidase P3, a new member of the TNF-TNFR2 signaling complex, induces phosphorylation of JNK1 and JNK2.
Aminopeptidase P3, a new member of the TNF-TNFR2 signaling complex, induces phosphorylation of JNK1 and JNK2.
Journal of cell science (2015-01-23)
Masaki Inoue, Haruhiko Kamada, Yasuhiro Abe, Kazuma Higashisaka, Kazuya Nagano, Yohei Mukai, Yasuo Yoshioka, Yasuo Tsutsumi, Shin-Ichi Tsunoda
ABSTRACT
Tumor necrosis factor (TNF) is an important mediator that triggers onset of autoimmune diseases and exerts its biological effects by interacting through two receptors, TNFR1 (also known as TNFRSF1A) and TNFR2 (also known as TNFRSF1B). TNFR2 signaling has significant potential to exert pro-survival and protective roles in several diseases. Unlike TNFR1 signaling, however, the mechanism of TNFR2 signal transduction is poorly understood, and few of its adaptor molecules are known. The present study utilized a proteomics approach to search for adaptor molecules in the TNFR2 signaling complex and identified aminopeptidase P3 (APP3, also known as XPNPEP3) to be a key molecule. One of its two isoforms, mitochondrial APP3 (APP3m) but not cytosolic APP3 (APP3c), was recruited to TNFR2 and shown to regulate TNF-TNFR2-dependent phosphorylation of JNK1 (also known as MAPK8) and JNK2 (also known as MAPK9). Furthermore, APP3m was released from mitochondria upon TNF stimulation in the absence of mitochondrial outer membrane permeabilization (MOMP). The observation of increased cell death upon downregulation of APP3m also suggested that APP3m exerts an anti-apoptotic function. These findings reveal that APP3m is a new member of the TNF-TNFR2 signaling complex and characterize an APP3-mediated TNFR2 signal transduction mechanism that induces activation of JNK1 and JNK2.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, suitable for electrophoresis, SDS-PAGE tested
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, ≥98% (NMR)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)