Merck
  • Stir bar sorptive extraction for the determination of pyrethroids in water samples A comparison between thermal desorption in a dedicated thermal desorber, in a split/splitless inlet and by liquid desorption.
Stir bar sorptive extraction for the determination of pyrethroids in water samples A comparison between thermal desorption in a dedicated thermal desorber, in a split/splitless inlet and by liquid desorption.
Journal of chromatography. A (2007-05-08)
Els Van Hoeck, Frank David, Pat Sandra
ABSTRACT
Stir bar sorptive extraction (SBSE) was applied to the enrichment of pyrethroids in water samples. Analysis was done by retention time-locked GC/MS in selected ion-monitoring (SIM) mode using trans-permethrin-d(6) as internal standard. Thermal desorption (TD) was performed in a classical split/splitless inlet equipped with a flip-top sealing system and in a dedicated thermal desorption unit. These two thermal desorption methods were compared to liquid desorption with ethyl acetate. Sensitivity was the highest for thermal desorption in the dedicated system and the lowest for liquid desorption even at injection volumes of 10 microL. The SBSE-TD-GC/MS method is able to determine pyrethroids at ultra-trace level (<1 ng/L) with RSD% smaller than 15 (n=6).
Product Description
Supelco
Inlet Liner, Split/Splitless Type, Straight Design (wool packed), pkg of 5 ea
Supelco
Inlet Liner, Split/Splitless Type, Straight Design (wool packed), pkg of 25 ea
Supelco
Inlet Liner, Splitless Type, Dual-Taper Design (unpacked), pkg of 5 ea
Supelco
Inlet Liner, Split Type, Cup Design (wool packed), pkg of 1 ea
Supelco
Inlet Liner, Splitless Type, Single Taper Design (unpacked), pkg of 25 ea
Supelco
Inlet Liner, Split/Splitless Type, Single Taper Design (wool packed), pkg of 5 ea
Supelco
Inlet Liner, Split Type, Cup Design (wool packed), pkg of 25 ea
Supelco
Inlet Liner, Split Type, Cup Design (unpacked), pkg of 5 ea
Supelco
Inlet Liner, Split/Splitless Type, Single Taper Design (wool packed), pkg of 25 ea
Supelco
Inlet Liner, Split Type, Cup Design (wool packed), pkg of 5 ea
Supelco
Inlet Liner, Splitless Type, Single Taper Design (unpacked), pkg of 5 ea