Merck
  • The role of the glutamatergic NMDA receptor in nanosilver-evoked neurotoxicity in primary cultures of cerebellar granule cells.
The role of the glutamatergic NMDA receptor in nanosilver-evoked neurotoxicity in primary cultures of cerebellar granule cells.
Toxicology (2013-12-03)
Elżbieta Ziemińska, Aleksandra Stafiej, Lidia Strużyńska
ABSTRACT
Nanoparticles are known to enter the vertebrate brain, but little is known about their neurotoxicity. The aim of this study is to investigate mechanisms of the contribution of AgNPs to neuronal cell death using primary cultures of rat cerebellar granule cells (CGCs). We tested the role of glutamatergic N-methyl-d-aspartate receptors (NMDA) in AgNP-evoked neurotoxicity using MK-801, a noncompetitive inhibitor of NMDAR. We used commercially available 0.2% PVP-coated AgNPs <100 nm in a concentration range of 2.5-75 μg/ml sonicated with fetal calf serum. After a 10 min incubation period, a dose-dependent increase in the uptake of (45)Ca(2+) into neurons was observed in the presence of 25-75 μg/mL AgNPs which was completely abolished by addition of MK-801. Using the fluorescent dye fluo3 AM we observed an increase in the intracellular calcium level by 87% compared to control. ROS production was found to increase by about 30% over control after a 30-min incubation with 75 μg/mL AgNPs. Further, we observed a significant decrease in the mitochondrial potential during a 30-min incubation with AgNPs. Administration of MK-801 was found to provide a protective effect. Our results show that excitotoxicity via activation of NMDA receptor, followed by calcium imbalance, destabilization of mitochondrial function and ROS production, indicate an important mechanism involved in neurotoxicity evoked by AgNPs in cultured neurons.
Sigma-Aldrich
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%
Sigma-Aldrich
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 2.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, granular, >250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver nanowires, diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver nanowires, diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, powder, <250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wool, diam. 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, ≥99.99% trace metals basis