Merck
  • Tetrabutylammonium 2-pyridyltriolborate salts for Suzuki-Miyaura cross-coupling reactions with aryl chlorides.
Tetrabutylammonium 2-pyridyltriolborate salts for Suzuki-Miyaura cross-coupling reactions with aryl chlorides.
Organic letters (2013-08-21)
Shohei Sakashita, Miho Takizawa, Juugaku Sugai, Hajime Ito, Yasunori Yamamoto
ABSTRACT
Palladium-catalyzed Suzuki-Miyaura cross-coupling reactions of tetrabutylammonium 2-pyridyltriolborate salts with various aryl (heteroaryl) chlorides can produce the corresponding desired coupling products with good to excellent yields. These tetrabutylammonium salts are more reactive than the corresponding lithium salts. The coupling reactions with aryl chlorides progressed in the presence of PdCl2dcpp (3 mol %) and CuI/MeNHCH2CH2OH (20 mol %) in anhydrous DMF without bases.
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic solution, 1.0 M in H2O
Sigma-Aldrich
Tetrabutylammonium cyanide, 95%
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, 97%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
Supelco
Tetrabutylammonium perchlorate, for electrochemical analysis, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium perchlorate, ≥95.0% (T)
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 53.5-56.5% in H2O
Sigma-Aldrich
Tetrabutylammonium cyanide, technical, ≥80%
Supelco
Tetrabutylammonium hydroxide solution, ~40% in water, suitable for ion chromatography
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic, puriss., 99% (T)
Supelco
Tetrabutylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Tetrabutylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Tetrabutylammonium bisulfate solution, suitable for ion pair chromatography, LiChropur, concentrate, ampule
Supelco
Tetrabutylammonium iodide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium iodide, ≥99.0% (AT)
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 75 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium nitrate, 97%
Sigma-Aldrich
Tetrabutylammonium chloride, ≥97.0% (NT)
Sigma-Aldrich
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
Supelco
Tetrabutylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium bromide solution, 50 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium iodide, reagent grade, 98%
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 1.0 M in THF
Sigma-Aldrich
Tetrabutylammonium azide