Merck
Physiology and pathophysiology of organic acids in cerebrospinal fluid.
Journal of inherited metabolic disease (1993-01-01)
G F Hoffmann, W Meier-Augenstein, S Stöckler, R Surtees, D Rating, W L Nyhan
ABSTRACT
Concentrations of organic acids in cerebrospinal fluid (CSF) appear to be directly dependent upon their rate of production in the brain. There is evidence that the net release of short-chain monocarboxylic acids from the brain is a major route for removing these products of cerebral metabolism. Concentrations of organic acids in blood and CSF are largely independent of each other. Quantitative reference values for the concentrations of organic acids in CSF and plasma as well as ratios of individual organic acids between CSF and plasma were determined in 35 pairs of samples from paediatric patients. Over 25 organic acids were quantifiable in all or in the majority of CSF and/or plasma specimens (limit of detection 1 mumol/L). There were substantial differences in the CSF/plasma ratios between subgroups of organic acids. Metabolites related to fatty-acid oxidation were present in CSF in substantially less amounts than in plasma. Organic acids related to carbohydrate and energy metabolism and to amino acid degradation were present in CSF in the same amounts as or slightly smaller amounts than in plasma. Finally, some organic acids were found in substantially higher amounts in CSF than in plasma, e.g. glycolate, glycerate, 2,4-dihydroxybutyrate, citrate and isocitrate. Studies of organic acids in CSF and plasma samples are presented from patients with 'cerebral' lactic acidosis, disorders of propionate and methylmalonate metabolism, glutaryl-CoA dehydrogenase deficiency and L-2-hydroxy-glutaric aciduria. It became apparent that derangements of organic acids in the CSF may occur independently of the systemic metabolism. Quantitative organic acid analysis in CSF will yield new information on the pathophysiology in the central nervous system (CNS) of these disorders and may prove necessary for successful monitoring of treatment of organoacidopathies, which present mainly with neurological disease. For example, in glutaryl-CoA dehydrogenase deficiency the urinary excretion of glutarate appears to be an inadequate parameter for monitoring the effect of dietary therapy, without plasma and CSF determinations. In L-2-hydroxyglutaric aciduria the elevation of L-2-hydroxyglutarate was found to be greater in CSF than in plasma. In addition, some other organic acids, glycolate, glycerate, 2,4-dihydroxybutyrate, citrate and isocitrate, were also elevated in the CSF of the patients out of proportion to normal levels in plasma and urine. High concentrations of an unknown compound, which was tentatively identified as 2,4-dihydroxyglutarate, were found in the CSF of patients with L-2-hydroxyglutaric aciduria.(ABSTRACT TRUNCATED AT 400 WORDS)
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Malic acid, 99%
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
L-(−)-Malic acid, 97%, optical purity ee: 99% (GLC)
Sigma-Aldrich
DL-Malic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
DL-Malic acid, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)
Sigma-Aldrich
Succinic acid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Succinic acid, puriss. p.a., ACS reagent, ≥99.5% (T)
Sigma-Aldrich
Pyruvic acid, 95%
Sigma-Aldrich
Azelaic acid, technical, ~85% (GC)
Sigma-Aldrich
L-(+)-Lactic acid, 80%
Sigma-Aldrich
Suberic acid, purum, ≥98.0% (T)
Sigma-Aldrich
Fumaric acid, ≥99.0% (T)
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
L-(−)-Malic acid, ReagentPlus®, ≥99%
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Pyruvic acid, ≥97%, FG
Sigma-Aldrich
Pyruvic acid, natural, ≥95%, FG
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
2-Hydroxy-3-methylbutyric acid, 99%
Sigma-Aldrich
Methylmalonic acid, 99%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Suberic acid, 98%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Fumaric acid, FCC, FG
Sigma-Aldrich
Azelaic acid, 98%
Sigma-Aldrich
Succinic acid, ACS reagent, ≥99.0%