Merck
  • Effect of three different contamination removal methods on bond strength of a self-etching adhesive to dentin contaminated with an aluminum chloride hemostatic agent.
Effect of three different contamination removal methods on bond strength of a self-etching adhesive to dentin contaminated with an aluminum chloride hemostatic agent.
The journal of contemporary dental practice (2013-04-13)
Amir Ahmad Ajami, Mehdi Abed Kahnamoii, Soodabeh Kimyai, Siavash Savadi Oskoee, Fatemeh Pournaghi-Azar, Mahmoud Bahari, Maryam Firouzmandi
ABSTRACT
This study evaluated the effect of three different contamination removal methods on bond strength of one-step self-etching adhesive to dentin contaminated with an aluminum chloride hemostatic agent. One hundred noncarious adult molars were used in this study; 2-mm-thick dentin disks were prepared and randomly assigned to 5 groups (n = 20). Group I: control; group II: hemostatic agent-contaminated; group III: hemostatic agent-contaminated and rinsing with water; group IV: hemostatic agent-contaminated and ethylene diamine tetraacetic acid (EDTA) application; and group V: hemostatic agent-contaminated and phosphoric acid application. Clearfil S3 Bond was used to bond composite to dentin surfaces. Subsequent to adding composite cylinders the shear bond strength test was performed. Data were analyzed by one-way ANOVA and Tukey test. Two additional specimens from each group were prepared and evaluated under scanning electron microscope (SEM). There were statistically significant differences in bond strength among the groups (p < 0.001). In two-by-two comparisons statistically significant differences were observed in bond strength values between all the groups (p < 0.001) except for groups I and IV (p = 0.933). Aluminum chloride hemostatic agent adversely affected the bond strength of self-etch adhesive to dentin. However, application of EDTA increased the bond strength to the level of normal dentin. EDTA and phosphoric acid removed aluminum chloride hemostatic agent contamination of dentin surfaces. However, unlike EDTA phosphoric acid failed to increase the bond strength of self-etch adhesive to dentin to the level of the control group.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, 98.5-101.5%, BioUltra
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aluminum chloride, reagent grade, 98%
Sigma-Aldrich
Aluminum chloride solution, 1.0 M in nitrobenzene
Sigma-Aldrich
Aluminum chloride, ReagentPlus®, 99%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Aluminum chloride hydrate, 99.999% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder