Skip to Content
Merck
All Photos(1)

Key Documents

03677

Sigma-Aldrich

Ethylenediaminetetraacetic acid disodium salt dihydrate

Ethylenediaminetetraacetic acid disodium salt dihydrate

BioUltra, for molecular biology, ≥99.0% (T)

Synonym(s):

Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€98.70
500 G
€310.00
1 KG
€554.00

€98.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€98.70
500 G
€310.00
1 KG
€554.00

About This Item

Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
CAS Number:
Molecular Weight:
372.24
Beilstein:
3900609
EC Number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.25
grade:
Molecular Biology
for molecular biology
form:
powder or crystals
solubility:
H2O: 0.1 M at 20 °C, clear, colorless

€98.70


Please contact Customer Service for Availability

Request a Bulk Order

grade

Molecular Biology
for molecular biology

Quality Level

product line

BioUltra

Assay

≥99.0% (T)

form

powder or crystals

reaction suitability

reagent type: chelator

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

pH

4.0-5.5 (25 °C, 0.1 M in H2O)

mp

248 °C (dec.) (lit.)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.1 M in H2O

UV absorption

λ: 260 nm Amax: 0.145
λ: 280 nm Amax: 0.025

SMILES string

O.O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O.OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2

InChI key

OVBJJZOQPCKUOR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Chelator of divalent cations. Inhibits enzymes, such as metalloproteases, that require divalent cations for activity.
Ethylenediaminetetraacetic acid disodium salt dehydrate can be used to study molecular biology, biochemicals, solutions and reagents, biological buffers, DNA and RNA purification, inorganic salts, essential chemicals and chelators. Ethylenediaminetetraacetic acid disodium salt dehydrate has been used in a study to advance monitoring of illegal thyreostatic drugs in livestock. Ethylenediaminetetraacetic acid disodium salt dehydrate has also been used in a study for the determination of 232Th in seawater by ICP-MS after preconcentration and separation using a chelating resin.

Other Notes

Running-buffer component for non-denaturing or urea-containing denaturing polyacrylamide gels in nucleic acid electrophoresis[1]

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R.C. Ogden et al.
Methods in Enzymology, 152, 65-65 (1987)
J Vanden Bussche et al.
Analytical and bioanalytical chemistry, 403(10), 2973-2982 (2012-02-22)
Thyreostatic drugs, illegally administrated to livestock for fattening purposes, are banned in the European Union since 1981. For monitoring their illegal use, sensitive and specific analytical methods are required. In this context, the knowledge of the stability in a matrix
G Lima-Oliveira et al.
British journal of biomedical science, 70(1), 6-9 (2013-04-27)
This study assesses the use of different dry K2 (dipotassium) EDTA vacuum tubes and whether or not they might represent a bias in haematological testing. Blood was collected in three dipotassium EDTA vacuum tubes from different manufacturers: Venosafe, Vacuette and
Suli Huang et al.
PloS one, 10(2), e0117007-e0117007 (2015-02-07)
microRNA (miRNA) plays a role in the pathogenesis of ischemic stroke, and single nucleotide polymorphisms in miRNA genes may contribute to disease susceptibility. However, the effect of miR-146a, miR-196a2, and miR-499 polymorphisms on ischemic stroke susceptibility has been rarely reported.
Lisa M Ryno et al.
Biomacromolecules, 15(8), 2944-2951 (2014-07-18)
End-functionalized macromolecular starch reagents, prepared by reductive amination, were grafted onto a urethane-linked polyester-based backbone using copper-catalyzed azide-alkyne cycloaddition (CuAAC) chemistry to produce novel amphiphilic hybrid graft copolymers. These copolymers represent the first examples of materials where the pendant chains

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service