Merck
  • Simple cyclohexanediamine-derived primary amine thiourea catalyzed highly enantioselective conjugate addition of nitroalkanes to enones.
Simple cyclohexanediamine-derived primary amine thiourea catalyzed highly enantioselective conjugate addition of nitroalkanes to enones.
Organic letters (2009-06-03)
Kui Mei, Mei Jin, Shilei Zhang, Ping Li, Wenjing Liu, Xiaobei Chen, Fei Xue, Wenhu Duan, Wei Wang
ABSTRACT
A highly enantioselective conjugate addition of nitroalkanes to enones has been developed. The process is efficiently catalyzed by a simple chiral cyclohexanediamine-derived primary amine thiourea with a broad substrate scope.
Sigma-Aldrich
(1R,2R)-(−)-1,2-Diaminocyclohexane, 98%
Sigma-Aldrich
(±)-trans-1,2-Diaminocyclohexane, 99%
Sigma-Aldrich
(1S,2S)-(+)-1,2-Diaminocyclohexane, 98%
Sigma-Aldrich
1,2-Diaminocyclohexane, mixture of cis and trans, 99%