Merck
  • Preparation of ethyl magnesium bromide for regiospecific analysis of triacylglycerols.
Preparation of ethyl magnesium bromide for regiospecific analysis of triacylglycerols.
Journal of oleo science (2008-07-16)
Yasuhiro Ando, Yuki Tomita, Yusuke Haba
ABSTRACT
This paper presents a procedure for preparation of a Grignard reagent, ethyl magnesium bromide, used for partial deacylation of triacylglycerols (TAG) in their regiospecific analysis. Magnesium turnings were reacted with ethereal solution of bromoethane in a screw-capped test tube to synthesize 2 mL of 1 M ethyl magnesium bromide. Continuously stirred with a vortex mixer, the reaction smoothly proceeded at room temperature. Regiospecific analysis of 1,3-distearoyl-2-oleoylglycerol using this product showed that fatty acid compositions of the sn-1(3) and sn-2 positions were contaminated by less than 2 mol% of fatty acids migrated from isomeric positions. The analyses of lard and cod liver/mackerel oil TAG showed typical distribution patterns of 16:0, 22:5n-3 and 22:6n-3 in pig and fish depot TAG. These results confirmed the view that the freshly prepared reagent is usable for regiospecific analysis of TAG.
Sigma-Aldrich
Ethylmagnesium bromide solution, 3.4 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Ethylmagnesium bromide solution, 1.0 M in tert-butyl methyl ether
Sigma-Aldrich
Ethylmagnesium bromide solution, 3.0 M in diethyl ether
Sigma-Aldrich
Ethylmagnesium bromide solution, 40% in 2-methyltetrahydrofuran
Sigma-Aldrich
Ethylmagnesium bromide solution, 1.0 M in THF
Sigma-Aldrich
Bromoethane, reagent grade, 98%
Sigma-Aldrich
Bromoethane, ReagentPlus®, ≥99%