Merck
  • In silico activity profiling reveals the mechanism of action of antimalarials discovered in a high-throughput screen.
In silico activity profiling reveals the mechanism of action of antimalarials discovered in a high-throughput screen.
Proceedings of the National Academy of Sciences of the United States of America (2008-06-27)
David Plouffe, Achim Brinker, Case McNamara, Kerstin Henson, Nobutaka Kato, Kelli Kuhen, Advait Nagle, Francisco Adrián, Jason T Matzen, Paul Anderson, Tae-Gyu Nam, Nathanael S Gray, Arnab Chatterjee, Jeff Janes, S Frank Yan, Richard Trager, Jeremy S Caldwell, Peter G Schultz, Yingyao Zhou, Elizabeth A Winzeler
ABSTRACT
The growing resistance to current first-line antimalarial drugs represents a major health challenge. To facilitate the discovery of new antimalarials, we have implemented an efficient and robust high-throughput cell-based screen (1,536-well format) based on proliferation of Plasmodium falciparum (Pf) in erythrocytes. From a screen of approximately 1.7 million compounds, we identified a diverse collection of approximately 6,000 small molecules comprised of >530 distinct scaffolds, all of which show potent antimalarial activity (<1.25 microM). Most known antimalarials were identified in this screen, thus validating our approach. In addition, we identified many novel chemical scaffolds, which likely act through both known and novel pathways. We further show that in some cases the mechanism of action of these antimalarials can be determined by in silico compound activity profiling. This method uses large datasets from unrelated cellular and biochemical screens and the guilt-by-association principle to predict which cellular pathway and/or protein target is being inhibited by select compounds. In addition, the screening method has the potential to provide the malaria community with many new starting points for the development of biological probes and drugs with novel antiparasitic activities.
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
O-Phospho-L-serine
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Neocuproine, ≥98%
Sigma-Aldrich
Anisomycin from Streptomyces griseolus, ≥98% (HPLC), solid
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
2-[4-(Dimethylamino)styryl]-1-ethylpyridinium iodide, ≥99% (HPLC), solid
Sigma-Aldrich
Cinchonine, crystallized, ≥98.0% (NT)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Bathophenanthroline, 97%
Sigma-Aldrich
Crystal Violet, S. No.: 785
Sigma-Aldrich
3,3′-Diethylthiacarbocyanine iodide, Dye content 95 %
Sigma-Aldrich
(+)-Cinchonine, 85%
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Crystal Violet, certified by the BSC
Sigma-Aldrich
Clotrimazole
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
9,10-Dimethylanthracene, 99%
Sigma-Aldrich
Crystal Violet
Sigma-Aldrich
Gentian violet
Sigma-Aldrich
Berberine chloride form