Merck
  • The Retinal Ganglion Cell Transportome Identifies Proteins Transported to Axons and Presynaptic Compartments in the Visual System In Vivo.
The Retinal Ganglion Cell Transportome Identifies Proteins Transported to Axons and Presynaptic Compartments in the Visual System In Vivo.
Cell reports (2019-08-15)
Lucio M Schiapparelli, Sahil H Shah, Yuanhui Ma, Daniel B McClatchy, Pranav Sharma, Jianli Li, John R Yates, Jeffrey L Goldberg, Hollis T Cline
ABSTRACT
The brain processes information and generates cognitive and motor outputs through functions of spatially organized proteins in different types of neurons. More complete knowledge of proteins and their distributions within neuronal compartments in intact circuits would help in the understanding of brain function. We used unbiased in vivo protein labeling with intravitreal NHS-biotin for discovery and analysis of endogenous axonally transported proteins in the visual system using tandem mass spectrometric proteomics, biochemistry, and both light and electron microscopy. Purification and proteomic analysis of biotinylated peptides identified ∼1,000 proteins transported from retinal ganglion cells into the optic nerve and ∼575 biotinylated proteins recovered from presynaptic compartments of lateral geniculate nucleus and superior colliculus. Approximately 360 biotinylated proteins were differentially detected in the two retinal targets. This study characterizes axonally transported proteins in the healthy adult visual system by analyzing proteomes from multiple compartments of retinal ganglion cell projections in the intact brain.
Sigma-Aldrich
Anti-Tau antibody, Mouse monoclonal, clone Tau46, purified from hybridoma cell culture
Sigma-Aldrich
Anti-MAP2 Antibody, clone AP20, clone AP20, Chemicon®, from mouse
Sigma-Aldrich
Anti-Synaptotagmin I Antibody, Chemicon®, from mouse
Sigma-Aldrich
Anti-Munc-18-1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-AIP1/Alix Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture