Merck
  • Expression of the monocyte chemotactic protein-1-induced protein 1 decreases human neuroblastoma cell survival.
Expression of the monocyte chemotactic protein-1-induced protein 1 decreases human neuroblastoma cell survival.
Oncology reports (2014-03-15)
Anna Skalniak, Elżbieta Boratyn, Sylwia D Tyrkalska, Irena Horwacik, Małgorzata Durbas, Maria Lastowska, Jolanta Jura, Hanna Rokita
ABSTRACT
The importance of monocyte chemotactic protein-1-induced protein 1 (MCPIP1) in the negative regulation of inflammatory reactions has already been extensively studied. However, its role in cancer is not yet established. We studied MCPIP1 gene expression in primary human neuroblastomas and several neuroblastoma cell lines. Our results showed a lack of MCPIP1 expression in primary neuroblastoma tumors. Moreover, it was found that the low expression of the protein measured in human neuroblastoma cell lines might be important for neuroblastoma survival, since enforced MCPIP1 gene expression in human neuroblastoma BE(2)-C cells caused a significant decrease in neuroblastoma cell viability and proliferation.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-ZC3H12A antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Roche
Cell Proliferation ELISA, BrdU (colorimetric), sufficient for ≤1,000 tests