Skip to Content
BE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
NaCl Solution (phys.)
Products
Product Category
buffers (2)
media (1)
Brand
Millipore (3)
Color
Physical Form
Agency
Chemical Composition
Capacity (mL)
Volume (ml)
Quality Segment
Closure Type
Application
Package Size
Sterility
Sterilization Method
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
1.46569
NaCl Solution (phys.),
bottle of, ready-to-use, bottle volume 100 mL , filling volume
Expand
View Pricing
1.46217
NaCl Solution (phys.),
ready-to-use, tube volume 10 mL , Filling Volume
Expand
View Pricing
1.46193
NaCl Solution (phys.),
ready-to-use, tube volume 9 mL , Filling Volume
Expand
View Pricing