Skip to Content
Merck
Emodin analytical standard

Emodin

Emodin analytical standard
Synonym(s):
1,3,8-Trihydroxy-6-methyl-9,10-anthracenedione, 1,3,8-Tri­hydroxy-6-methyl­anthra­quinone, 6-Methyl-1,3,8-tri­hydroxy­anthra­quinone, Archin, Emodol, Frangula-emodin, Frangulic acid, Rheum emodin, Emodin
Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique