Skip to Content
Merck
2-(2-Aminoethoxy)ethanol 98%

2-(2-Aminoethoxy)ethanol

2-(2-Aminoethoxy)ethanol 98%
Synonym(s):
Diethylene glycolamine, 2-(2-Aminoethoxy)ethanol
Linear Formula:
NH2CH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
105.14

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing