Skip to Content
Merck
All Photos(2)

Key Documents

8.18431

Sigma-Aldrich

2-(2-Aminoethoxy)ethanol

for synthesis

Synonym(s):

2-(2-Aminoethoxy)ethanol, Ethylene glycol mono(2-aminoethyl) ether

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€42.80
500 ML
€74.70

€42.80


Estimated to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€42.80
500 ML
€74.70

About This Item

Empirical Formula (Hill Notation):
C4H11NO2
CAS Number:
Molecular Weight:
105.14
MDL number:
UNSPSC Code:
12352104
EC Index Number:
213-195-4
NACRES:
NA.22

€42.80


Estimated to ship on02 April 2025Details


Request a Bulk Order

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

370 °C

potency

3000 mg/kg LD50, oral (Rat)
>3000 mg/kg LD50, skin (Rabbit)

expl. lim.

2.0-15.5 % (v/v)

pH

10.2 (20 °C, 10 g/L in H2O)

bp

218-224 °C/1013 mbar

mp

-11 °C

transition temp

flash point 127 °C

density

1.06 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

NCCOCCO

InChI

1S/C4H11NO2/c5-1-3-7-4-2-6/h6H,1-5H2

InChI key

GIAFURWZWWWBQT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.054 - 1.055
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

260.6 °F

Flash Point(C)

127 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service