Skip to Content
Merck
All Photos(1)

Key Documents

V900886

Sigma-Aldrich

Select Yeast Extract

Vetec, reagent grade, powder, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352205
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

form

powder

technique(s)

cell culture | mammalian: suitable

storage temp.

room temp

SMILES string

OC(=O)CC1c2c3c(ccc2c4c1cccc4)cccc3

InChI

1S/C19H14O2/c20-18(21)11-17-15-8-4-3-7-14(15)16-10-9-12-5-1-2-6-13(12)19(16)17/h1-10,17H,11H2,(H,20,21)

InChI key

GQNBDGXKDJSVGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For general bacteriological use with a variety of microorganisms.
Yeast extract is used as a bacteriological media component for a variety of microorganisms and molecular genetics applications.

Other Notes

Water soluble portion of autolyzed yeast with intact B-complex vitamins. Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates and can be used as additive for culture media.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service