Skip to Content
Merck
All Photos(1)

Documents

V900803

Sigma-Aldrich

Formic acid

Vetec, reagent grade, 95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOOH
CAS Number:
Molecular Weight:
46.03
Beilstein:
1209246
EC Number:
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:

grade

reagent grade

vapor density

1.6 (vs air)

vapor pressure

44.8 mmHg ( 20 °C)

product line

Vetec

Assay

95%

form

liquid

autoignition temp.

1004 °F

expl. lim.

57 %

refractive index

n20/D 1.370 (lit.)

pH

2.2 (20 °C, 2.2 g/L)

bp

100-101 °C (lit.)

mp

8.2-8.4 °C (lit.)

density

1.22 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OC=O

InChI

1S/CH2O2/c2-1-3/h1H,(H,2,3)

InChI key

BDAGIHXWWSANSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

121.1 °F - closed cup

Flash Point(C)

49.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emma Granqvist et al.
The New phytologist, 207(3), 551-558 (2015-05-27)
Plants that form root-nodule symbioses are within a monophyletic 'nitrogen-fixing' clade and associated signalling processes are shared with the arbuscular mycorrhizal symbiosis. Central to symbiotic signalling are nuclear-associated oscillations in calcium ions (Ca(2+) ), occurring in the root hairs of
J Liesivuori et al.
Pharmacology & toxicology, 69(3), 157-163 (1991-09-01)
Metabolism of methanol, methyl ethers, esters and amides give rise to formic acid. This acid is an inhibitor of the mitochondrial cytochrome oxidase causing histotoxic hypoxia. Formic acid is a weaker inhibitor than cyanide and hydrosulphide anions. The body burden
Tasqiah Julianti et al.
Journal of ethnopharmacology, 155(1), 426-434 (2014-06-04)
Leaf decoctions of Carica papaya have been traditionally used in some parts of Indonesia to treat and prevent malaria. Leaf extracts and fraction have been previously shown to possess antiplasmodial activity in vitro and in vivo. Antiplasmodial activity of extracts
Yoann Rombouts et al.
Annals of the rheumatic diseases, 74(1), 234-241 (2013-10-10)
To determine whether anticitrullinated protein antibodies (ACPA) exhibit specific changes in Fc glycosylation prior to the onset of arthritis. Serum samples of patients with ACPA-positive arthralgia (n=183) were collected at baseline and at various time points of follow-up. 105 patients
S Baron-Cohen et al.
Molecular psychiatry, 20(3), 369-376 (2014-06-04)
Autism affects males more than females, giving rise to the idea that the influence of steroid hormones on early fetal brain development may be one important early biological risk factor. Utilizing the Danish Historic Birth Cohort and Danish Psychiatric Central

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service