Skip to Content
Merck
All Photos(1)

Key Documents

V900571

Sigma-Aldrich

1,10-Decanediol

Vetec, reagent grade, 98%

Synonym(s):

Decamethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)10OH
CAS Number:
Molecular Weight:
174.28
Beilstein:
1698975
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

SMILES string

OCCCCCCCCCCO

InChI

1S/C10H22O2/c11-9-7-5-3-1-2-4-6-8-10-12/h11-12H,1-10H2

InChI key

FOTKYAAJKYLFFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

305.6 °F - closed cup

Flash Point(C)

152 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Schwach-Abdellaoui et al.
Journal of biomaterials science. Polymer edition, 10(3), 375-389 (1999-04-03)
The synthesis of poly(ortho-esters) (POEs) containing lactic acid dimers in the polymer backbone for possible use in controlled drug release applications is described. These autocatalyzed POEs are prepared by the acid catalyzed condensation of 3,9-diethylidene-2,4,8,10-tetraoxaspiro(5,5)undecane with diols to produce linear
O A Vinogradova et al.
Bioorganicheskaia khimiia, 35(3), 384-396 (2009-07-22)
An opportunity of designing nontypical double-stranded DNA structures containing nonnatural inserts in a regular nucleotide DNA sequence has been investigated. The looped nucleotide inserts on the basis of adenylates and thymidilates, and nonnucleotide inserts on the basis of phosphodiesters of
J Helle et al.
Biomaterials, 23(22), 4397-4404 (2002-09-11)
Poly(ortho esters) with a low glass transition temperature are semi-solid materials so that therapeutic agents can be incorporated at room temperature, without the use of solvents, by a simple mixing procedure. When molecular weights are limited to < 5 kDa

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service