Skip to Content
Merck
All Photos(2)

Key Documents

V900560

Sigma-Aldrich

Pimelic acid

Vetec, reagent grade, 98%

Synonym(s):

Heptanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C(CH2)5CO2H
CAS Number:
Molecular Weight:
160.17
Beilstein:
1210024
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

bp

212 °C/10 mmHg (lit.)

mp

103-105 °C (lit.)

SMILES string

OC(=O)CCCCCC(O)=O

InChI

1S/C7H12O4/c8-6(9)4-2-1-3-5-7(10)11/h1-5H2,(H,8,9)(H,10,11)

InChI key

WLJVNTCWHIRURA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Niwa et al.
Clinica chimica acta; international journal of clinical chemistry, 99(1), 71-83 (1979-11-15)
(1) 2,4-Dimethyladipic acid was first identified in normal human urine using gas chromatography-mass spectrometry. Urinary excretion of 2,4-dimethyladipic acid in 7 healthy adults ranged from 4.9 mumol to 14 mumol per 24 h. (2) Succinic acid, adipic acid, 3-methyladipic acid
B R Pettit
Clinica chimica acta; international journal of clinical chemistry, 156(1), 85-90 (1986-04-15)
A selected ion monitoring assay for thiodiglycollic acid in urine is described. Urine samples are analysed by combined gas chromatography-mass spectrometry as their dibutyl esters using pimelic acid as an internal standard. Rapid analysis was achieved by the simplification of
V G Mahadevappa et al.
Biochimica et biophysica acta, 713(1), 73-79 (1982-10-14)
The molecular species composition of the individual diacyl phospholipids was determined in human platelets. The 1-acyl (16:0, 18:0, etc.) homologues of the various 2-acyl (16:0, 18:1, 18:2, 20:4, etc) species of the phosphatidylcholine, phosphatidylethanolamine, phosphatidylserine and phosphatidylinositol were assessed by
S Passi et al.
Acta dermato-venereologica. Supplementum, 143, 8-13 (1989-01-01)
Medium chain length dicarboxylic acids (DA) from C8 to C13 are competitive inhibitors of tyrosinase in vitro. The introduction of electron acceptor groups or electron donor groups into the 2 and/or the 8 position of the molecule enhances or reduces
C D Capo-Chichi et al.
The American journal of clinical nutrition, 71(4), 978-986 (2000-03-25)
Riboflavin, flavin mononucleotide (FMN), and flavin adenine dinucleotide (FAD) concentrations have been little studied in cases of malnutrition. Our objective was to investigate the effects of malnutrition on riboflavin status and riboflavin's relation with thyroid hormones and concentrations of urinary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service