Skip to Content
Merck
All Photos(1)

Key Documents

V900543

Sigma-Aldrich

Geraniol

Vetec, reagent grade, 97%

Synonym(s):

trans-3,7-Dimethyl-2,6-octadien-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2OH
CAS Number:
Molecular Weight:
154.25
Beilstein:
1722456
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

vapor density

5.31 (vs air)

product line

Vetec

Assay

97%

refractive index

n20/D 1.474 (lit.)

bp

229-230 °C (lit.)

density

0.879 g/mL at 20 °C (lit.)

SMILES string

C\C(C)=C\CC\C(C)=C\CO

InChI

1S/C10H18O/c1-9(2)5-4-6-10(3)7-8-11/h5,7,11H,4,6,8H2,1-3H3/b10-7+

InChI key

GLZPCOQZEFWAFX-JXMROGBWSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Lapczynski et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46 Suppl 11, S160-S170 (2008-07-22)
A toxicologic and dermatologic review of geraniol when used as a fragrance ingredient is presented.
Gavin L Sacks et al.
Journal of agricultural and food chemistry, 60(12), 2998-3004 (2012-03-09)
1,1,6-Trimethyl-1,2-dihydronaphthalene (TDN) is well-known to contribute "petrol" aromas to aged Riesling wines, but its prevalence and contribution to young Riesling or non-Riesling wines is not well understood. TDN concentrations were measured in 1-3-year-old varietal wines produced from Cabernet franc (n
Lina Hagvall et al.
Contact dermatitis, 68(4), 224-231 (2013-03-21)
Geraniol is a commonly used fragrance terpene, and is tested in the baseline series in fragrance mix I. Geraniol is a pro-hapten and a pre-hapten, and sensitizers are formed in the autoxidation and skin metabolism of geraniol. Previous patch testing
Edita E Revay et al.
Acta tropica, 124(1), 102-105 (2012-07-04)
We conducted a study to determine the degree of personal protection provided by the Terminix(®) ALLCLEAR(®) Mosquito Mister - Lantern Edition. This outdoor unit was operated to disperse an aerosolized aqueous 0.3% geraniol emulsion in timed-release intervals of 5.0, 7.5
Alex Van Moerkercke et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(26), 8130-8135 (2015-06-17)
Plants make specialized bioactive metabolites to defend themselves against attackers. The conserved control mechanisms are based on transcriptional activation of the respective plant species-specific biosynthetic pathways by the phytohormone jasmonate. Knowledge of the transcription factors involved, particularly in terpenoid biosynthesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service