Skip to Content
Merck
All Photos(2)

Documents

V900389

Sigma-Aldrich

D-(−)-Ribose

Vetec, reagent grade, 99%

Synonym(s):

Aldehydo-D-ribose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
Beilstein:
1723081
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

99%

form

powder

mp

88-92 °C (lit.)

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@@H](O)C([H])=O

InChI

1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h1,3-5,7-10H,2H2/t3-,4+,5-/m0/s1

InChI key

PYMYPHUHKUWMLA-LMVFSUKVSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Conditions and Mechanism of Formation of the Maillard Reaction Pigment, Furpenthiazinate, in a Model System and in Some Acid Hydrolyzates of Foods and its Biological Properties.: This article investigates the conditions and mechanisms under which ᴅ-(−)-Ribose participates in the Maillard reaction to form bioactive pigments like Furpenthiazinate. It details the implications for food chemistry and the nutritional properties of processed foods (Noda et al., 2024).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raman K Sharma et al.
Bioorganic & medicinal chemistry, 20(23), 6821-6830 (2012-10-27)
A series of peracetylated O-aryl α,β-d-ribofuranosides have been synthesized and an efficient biocatalytic methodology has been developed for the separation of their anomers which was otherwise almost impossible by column chromatographic or other techniques. The incubation of 2,3,5-tri-O-acetyl-1-O-aryl-α,β-d-ribofuranoside with Lipozyme®
Paul O Hassa et al.
Frontiers in bioscience : a journal and virtual library, 13, 3046-3082 (2007-11-06)
Poly-ADP-ribose metabolism plays a mayor role in a wide range of biological processes, such as maintenance of genomic stability, transcriptional regulation, energy metabolism and cell death. Poly-ADP-ribose polymerases (PARPs) are an ancient family of enzymes, as evidenced by the poly-ADP-ribosylating
Luigi J Alvarado et al.
Chembiochem : a European journal of chemical biology, 15(11), 1573-1577 (2014-06-24)
Isotope labeling has revolutionized NMR studies of small nucleic acids, but to extend this technology to larger RNAs, site-specific labeling tools to expedite NMR structural and dynamics studies are required. Using enzymes from the pentose phosphate pathway, we coupled chemically
Anders Virtanen et al.
Critical reviews in biochemistry and molecular biology, 48(2), 192-209 (2013-03-19)
Deadenylation of eukaryotic mRNA is a mechanism critical for mRNA function by influencing mRNA turnover and efficiency of protein synthesis. Here, we review poly(A)-specific ribonuclease (PARN), which is one of the biochemically best characterized deadenylases. PARN is unique among the
Carine Baraguey et al.
Organic & biomolecular chemistry, 11(16), 2638-2647 (2013-03-05)
The pivaloyloxymethyl (PivOM) group is a biolabile 2'-O-ribose protection that is under development in a prodrug-based approach for siRNA applications. Besides an expected cellular uptake, nucleic acid sequences carrying PivOM showed also increased nuclease resistance and, in most cases, an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service