Skip to Content
Merck
All Photos(1)

Key Documents

V900320

Sigma-Aldrich

Ethylhexadecyldimethylammonium bromide

Vetec, reagent grade, ≥98%

Synonym(s):

Cetyldimethylethylammonium bromide, Dimethylethylhexadecylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)15N+(CH3)2CH2CH3]Br-
CAS Number:
Molecular Weight:
378.47
Beilstein:
3729134
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥98%

mol wt

378.47 g/mol

storage temp.

2-8°C

SMILES string

[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)CC

InChI

1S/C20H44N.BrH/c1-5-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(3,4)6-2;/h5-20H2,1-4H3;1H/q+1;/p-1

InChI key

VUFOSBDICLTFMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mounesha N Garaga et al.
Physical chemistry chemical physics : PCCP, 17(33), 21664-21682 (2015-08-01)
Boron heteroatom distributions are shown to be significantly different in two closely related layered borosilicates synthesized with subtly different alkylammonium surfactant species. The complicated order and disorder near framework boron sites in both borosilicates were characterized at the molecular level
Bing Wu et al.
Environmental science & technology, 49(9), 5760-5770 (2015-04-09)
The ability of engineered nanomaterials (NMs) to act as inhibitors of ATP-binding cassette (ABC) efflux transporters in embryos of white sea urchin (Lytechinus pictus) was studied. Nanocopper oxide (nano-CuO), nanozinc oxide (nano-ZnO), and their corresponding metal ions (CuSO4 and ZnSO4)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service