Skip to Content
Merck
All Photos(2)

Key Documents

V900203

Sigma-Aldrich

Magnesium oxide

Vetec, reagent grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgO
CAS Number:
Molecular Weight:
40.30
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
reagent grade
form:
powder
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

form

powder

concentration

≥55% Mg (EDTA titration)

mp

2852 °C (lit.)

SMILES string

O=[Mg]

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyan Sun et al.
BMC plant biology, 15, 116-116 (2015-05-11)
Heat stress is a critical threat to tall fescue in transitional and warm climate zones. Identification of association between molecular markers and heat tolerance-related functional traits would promote the efficient selection of heat tolerant tall fescue cultivars. Association analysis of
Peter Visser et al.
Faraday discussions, 180, 511-526 (2015-05-01)
Lithium carbonate and lithium oxalate were incorporated as leachable corrosion inhibitors in model organic coatings for the protection of AA2024-T3. The coated samples were artificially damaged with a scribe. It was found that the lithium-salts are able to leach from
Sanket A Deshmukh et al.
Physical chemistry chemical physics : PCCP, 14(44), 15593-15605 (2012-10-19)
Atomic scale characterization of the structure and dynamics of confined water molecules located near the metal oxide-aqueous interface is carried out using molecular dynamics simulations. Proximity effects on water molecules (H(2)O) near a magnesium oxide surface (MgO(100)) at room temperature
Magdalena Maciejewska et al.
Journal of chromatography. A, 1257, 141-148 (2012-08-22)
Inverse gas chromatography (IGC) was applied at infinite dilution to evaluate the surface properties of calcium and magnesium oxide nanoparticles and the effect of surface grafted unsaturated carboxylic acid on the nanopowder donor-acceptor characteristics. The dispersive components (γ(s)(D)) of the
Jian-Bin Shi et al.
Carbohydrate research, 359, 65-69 (2012-08-29)
This work describes the structural changes of bagasse hemicelluloses during the cooking process involving active oxygen (O(2) and H(2)O(2)) and solid alkali (MgO). The hemicelluloses obtained from the bagasse raw material, pulp, and yellow liquor were analyzed by high-performance anion-exchange

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service