Skip to Content
Merck
All Photos(3)

Key Documents

V900128

Sigma-Aldrich

Hydrazine dihydrochloride

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2 · 2HCl
CAS Number:
Molecular Weight:
104.97
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
reagent grade
form:
crystals or chunks
powder or solid

grade

reagent grade

product line

Vetec

Assay

98%

form

crystals or chunks
powder or solid

mp

200 °C (dec.) (lit.)

density

1.42 g/mL at 25 °C (lit.)

SMILES string

Cl[H].Cl[H].NN

InChI

1S/2ClH.H4N2/c;;1-2/h2*1H;1-2H2

InChI key

LIAWOTKNAVAKCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ranendu Sekhar Das et al.
Dalton transactions (Cambridge, England : 2003), 42(11), 4068-4080 (2013-01-24)
PVP capped platinum nano particles (PNP) of 5 nm diameter were prepared and characterized as homogeneous and of spherical nature. At physiological pH range (6.0-8.0), these PNP catalyze the deoxygenation of phenoxazine group containing resazurin (1) by hydrazine. The observed
Boran Kartal et al.
FEMS microbiology reviews, 37(3), 428-461 (2012-12-06)
Anaerobic ammonium-oxidizing (anammox) bacteria primarily grow by the oxidation of ammonium coupled to nitrite reduction, using CO2 as the sole carbon source. Although they were neglected for a long time, anammox bacteria are encountered in an enormous species (micro)diversity in
Weidong Qu et al.
Biosensors & bioelectronics, 42, 430-433 (2012-12-19)
Graphene-nickel nanoparticle hybrid was prepared by the one-step far infrared-assisted reduction of graphene oxide and nickel (II) ions using hydrazine. It was loaded on the surface of a magnetic electrode for electrochemical sensing. The feasibility and performance of the novel
Paul D Rainville et al.
Bioanalysis, 4(23), 2833-2842 (2012-12-12)
Accurate mass based LC-MS combined with statistical analysis is established as a core analytical technology for metabonomic studies. This is primarily due to the specificity, sensitivity and structural elucidation capabilities of the technology. The vast majority of these studies are
Erika Lattová et al.
Mass spectrometry reviews, 32(5), 366-385 (2013-01-25)
Over the last years, extensive studies have evaluated glycans from different biological samples and validated the importance of glycosylation as one of the most important post-translational modifications of proteins. Although a number of new methods for carbohydrate analysis have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service