Skip to Content
Merck
All Photos(3)

Key Documents

V900119

Sigma-Aldrich

Urea

Vetec, reagent grade, 99%

Synonym(s):

Carbamide, Carbonyldiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

99%

mp

132-135 °C (lit.)

density

1.335 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Urea is a chaotropic agent and is used for protein denaturation. It disturbs the hydrogen bonds in the secondary, tertiary and quaternary structure of proteins. Urea can also disturb hydrogen bonds present in DNA secondary structure.[1]

Application

Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheehan D.
Physical Biochemistry: Principles and Applications (2013)
L Li et al.
Cell death and differentiation, 22(7), 1081-1093 (2014-12-20)
P53 is critically important in preventing oncogenesis but its role in inflammation in general and in the function of inflammatory macrophages in particular is not clear. Here, we show that bone marrow-derived macrophages exhibit endogenous p53 activity, which is increased
A mechanically and electrically self-healing supercapacitor.
Hua Wang et al.
Advanced materials (Deerfield Beach, Fla.), 26(22), 3638-3643 (2014-02-21)
Marco Benevento et al.
Nature communications, 5, 5613-5613 (2014-12-11)
The ectopic expression of Oct4, Klf4, c-Myc and Sox2 (OKMS) transcription factors allows reprogramming of somatic cells into induced pluripotent stem cells (iPSCs). The reprogramming process, which involves a complex network of molecular events, is not yet fully characterized. Here
Annette Bahlinger et al.
Angewandte Chemie (International ed. in English), 53(33), 8779-8783 (2014-03-20)
β-Amino thioesters are important natural building blocks for the synthesis of numerous bioactive molecules. An organocatalyzed Mannich reaction was developed which provides direct and highly stereoselective access to acyclic β(2)- and β(2,3,3)-amino thioesters with adjacent tertiary and quaternary stereocenters. Mechanistic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service