Skip to Content
Merck
All Photos(3)

Key Documents

V900014

Sigma-Aldrich

Pyrogallol

Vetec, reagent grade

Synonym(s):

1,2,3-Trihydroxybenzene, 2,3-Dihydroxyphenol, Pyrogallic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(OH)3
CAS Number:
Molecular Weight:
126.11
Beilstein:
907431
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade:
reagent grade
bp:
309 °C (lit.)
vapor pressure:
10 mmHg ( 167.7 °C)

grade

reagent grade

vapor density

4.4 (vs air)

vapor pressure

10 mmHg ( 167.7 °C)

product line

Vetec

form

solid

dilution

(for analytical testing)

bp

309 °C (lit.)

mp

133-134 °C (lit.)

SMILES string

Oc1cccc(O)c1O

InChI

1S/C6H6O3/c7-4-2-1-3-5(8)6(4)9/h1-3,7-9H

InChI key

WQGWDDDVZFFDIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer C Chapin et al.
Journal of the American Chemical Society, 134(36), 15000-15009 (2012-09-04)
Pyrogallol[4]arene is a macrocycle with a concave surface and 12 peripheral hydroxyl groups that mediate its self-assembly to form hexamers of octahedral symmetry in the solid state, in solution, and in the gas phase. These hexamers enclose approximately 1300 Å(3)
M Buchweitz et al.
Food chemistry, 135(4), 3010-3019 (2012-09-18)
The formation of anthocyanin-metal chelates, exhibiting intense blue colours was monitored over a period up to 10 weeks. Evaluating normalised absorption spectra in the range of 580-700 nm and their proportion of the total area under the curve (AUC), provided
Mohammad Ghadiri et al.
Ecotoxicology and environmental safety, 91, 117-121 (2013-02-26)
This paper presents a new method for spectrophotometirc detection of sulfide applying fungal peroxidase immobilized on sodium alginate. The sensing scheme was based on decrease of the absorbance of the orange compound, purpurogallin produced from pyrogallol and H2O2 as substrates
Edgar Nagles et al.
Talanta, 99, 119-124 (2012-09-13)
An adsorptive stripping voltammetric (AdSV) method is presented for the simultaneous determination of Pb(II) and Cd(II) at trace levels in natural waters, based on metal complexation with pyrogallol red (PR) and subsequent adsorptive deposition on a Nafion-mercury coated glassy carbon
Jeannette Geschwind et al.
Macromolecular rapid communications, 34(2), 150-155 (2012-11-30)
A series of functional polycarbonates, poly((isopropylidene glyceryl glycidyl ether)-co-(glycidyl methyl ether) carbonate) (P((IGG-co-GME) C)) random copolymers with different fractions of 1,2-isopropylidene glyceryl glycidyl ether (IGG) units, is synthesized. After acidic hydrolysis of the acetal protecting groups, a new type of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service