Skip to Content
Merck
All Photos(1)

Key Documents

V800389

Sigma-Aldrich

Sodium molybdate

LR, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2MoO4
CAS Number:
Molecular Weight:
205.92
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
LR
form:
crystalline powder
Pricing and availability is not currently available.

grade

LR

product line

Vetec

Assay

≥99.5%

form

crystalline powder

pH

7.0-10.5

mp

687 °C (lit.)

density

3.78 g/mL at 25 °C (lit.)

SMILES string

[Na+].[Na+].[O-][Mo]([O-])(=O)=O

InChI

1S/Mo.2Na.4O/q;2*+1;;;2*-1

InChI key

TVXXNOYZHKPKGW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saraswathi Panneerselvam et al.
Chemico-biological interactions, 145(2), 159-163 (2003-04-11)
Diabetes mellitus is a disease, which virtually affects all the systems in the body including the immune system. Bacterial infections are important causes of increased morbidity and mortality in diabetic patients. In the present study, we assessed the effect of
Akira Sakakura et al.
Chemical communications (Cambridge, England), (30)(30), 3561-3563 (2008-07-26)
Efficient total syntheses of fluvibactin and vibriobactin have been achieved via molybdenum(VI) oxide-catalyzed dehydrative cyclization, Sb(OEt)(3)-catalyzed ester-amide transformation, and WSCI and HOAt-promoted dehydrative amide formation.
Laurent Caron et al.
Journal of colloid and interface science, 282(2), 478-485 (2004-12-14)
Chemically generated singlet oxygen (1O2, 1Deltag) is able to oxidize a great deal of hydrophobic substrates from molybdate-catalyzed hydrogen peroxide decomposition, provided a suitable reaction medium such as a microemulsion system is used. However, high substrate concentrations or poorly reactive
Lluís Palou et al.
Pest management science, 58(5), 459-466 (2002-05-10)
The effectiveness of low-toxicity chemicals as possible alternatives to synthetic fungicides for the control of post-harvest green and blue moulds of citrus was evaluated. A preliminary selection of chemicals, mostly common food additives, was made through in vivo primary screenings
Bo-Geon Yun et al.
Biochemistry, 43(25), 8217-8229 (2004-06-24)
Hsp90 functions to facilitate the folding of newly synthesized and denatured proteins. Hsp90 function is modulated through its interactions with cochaperones and the binding and hydrolysis of ATP. Recently, novobiocin has been shown to bind to a second nucleotide binding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service