Skip to Content
Merck
All Photos(1)

Key Documents

V800269

Sigma-Aldrich

N,N-Dimethylaniline

LR, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N(CH3)2
CAS Number:
Molecular Weight:
121.18
Beilstein:
507140
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade:
LR
bp:
193-194 °C (lit.)
Pricing and availability is not currently available.

grade

LR

product line

Vetec

Assay

≥99%

form

liquid

dilution

(for analytical testing)

refractive index

n20/D 1.557 (lit.)

pH

7.4 (20 °C, 1.2 g/L)

bp

193-194 °C (lit.)

mp

1.5-2.5 °C (lit.)

density

0.956 g/mL at 25 °C (lit.)

SMILES string

CN(C)c1ccccc1

InChI

1S/C8H11N/c1-9(2)8-6-4-3-5-7-8/h3-7H,1-2H3

InChI key

JLTDJTHDQAWBAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenji Hirai et al.
Chemical communications (Cambridge, England), 48(52), 6472-6474 (2012-04-21)
Spatiospecific functionalisation of a shell crystal was performed in a core-shell crystal of a porous coordination polymer (PCP) via post-synthetic modification (PSM). The shell crystal allowed the core crystal to selectively accumulate N,N-dimethylaniline (DMA) and afford the intense exciplex fluorescence.
Hohjai Lee et al.
Nano letters, 11(12), 5367-5372 (2011-11-03)
We used a fluorescent chemical indicator of magnetic field to visualize the magnetic field around ferromagnetic nanostructures. The indicator was a chain-linked electron donor-acceptor molecule, phenanthrene-(CH2)12-O-(CH2)2-dimethylaniline, that forms spin-correlated radical pairs upon photoexcitation. The magnetic field altered the coherent spin
Subashani Maniam et al.
Organic letters, 10(10), 1885-1888 (2008-04-16)
Cyclodextrin [2]rotaxanes have been prepared by coupling dimethylanilines with dicarboxylic acids using DMT-MM, in aqueous solutions of alpha-cyclodextrin, and the example illustrated shows unusual fluorescence emission and other spectroscopic behavior characteristic of the formation of molecular wires in solution, similar
Atanu Kumar Das et al.
The journal of physical chemistry. B, 115(16), 4680-4688 (2011-04-07)
Ultrafast photoinduced electron transfer (PET) from N,N-dimethylaniline (DMA) to coumarin dyes in a room-temperature ionic liquid (RTIL, [pmim][BF(4)]) and in a mixed micelle containing the RTIL and a triblock copolymer, (PEO)(20)-(PPO)(70)-(PEO)(20), (Pluronic P123) is studied using femtosecond upconversion. A Marcus-like
Zhiwei Lin et al.
Journal of the American Chemical Society, 131(50), 18060-18062 (2009-11-26)
Ultrafast UV-vibrational spectroscopy was used to investigate how vibrational excitation of the bridge changes photoinduced electron transfer between donor (dimethylaniline) and acceptor (anthracene) moieties bridged by a guanosine-cytidine base pair (GC). The charge-separated (CS) state yield is found to be

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service