Skip to Content
Merck
All Photos(1)

Key Documents

V800250

Sigma-Aldrich

Manganese(II) sulfate monohydrate

AR, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnSO4 · H2O
CAS Number:
Molecular Weight:
169.02
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

AR

product line

Vetec

Assay

≥99%

SMILES string

O.[Mn++].[O-]S([O-])(=O)=O

InChI

1S/Mn.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

ISPYRSDWRDQNSW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edson R Silva et al.
Experimental parasitology, 125(2), 152-155 (2010-01-30)
In Leishmania, arginase is responsible for the production of ornithine, a precursor of polyamines required for proliferation of the parasite. In this work, the activation kinetics of immobilized arginase enzyme from L. (L.) amazonensis were studied by varying the concentration
Melanie F Struve et al.
American journal of industrial medicine, 50(10), 772-778 (2007-07-11)
Manganese neurotoxicity in humans is recognized as a form of parkinsonism with lesions occurring predominantly within the globus pallidus, subthalamic nucleus, putamen, and caudate nucleus. This study evaluated dopamine, 3,4-dihydroxyphenylacetic acid, homovanillic acid, serotonin, norepinephrine, 5-hydroxyindoleacetic acid, gamma-aminobutyric acid (GABA)
Haixing Li et al.
Amino acids, 38(5), 1439-1445 (2009-09-30)
Production of gamma-aminobutyric acid (GABA) was carried out in Erlenmeyer flasks by Lactobacillus brevis NCL912. Traditional methods were first adopted to select the key factors that impact the GABA production to preliminarily determine the suitable concentration ranges of the key
Xiaojun Yang et al.
Animal science journal = Nihon chikusan Gakkaiho, 83(6), 474-481 (2012-06-15)
Iron is often found to be of excessive concentrations in laying hens' diets, which may cause antagonistic interactions with other minerals. This study was conducted to investigate how to supplement Zn and Mn in the diets without Fe supplementation. In
Long Peng et al.
Journal of colloid and interface science, 377(1), 13-17 (2012-04-21)
A facile single-source precursor method has been applied for the selective synthesis of MnS nanocrystals (NCs) with well-defined shapes and crystal structures such as hexapod, octahedral, hexagonal shaped α-MnS NCs, and pencil-shaped γ-MnS NCs. The effects of the composition of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service