Skip to Content
Merck
All Photos(1)

Key Documents

V800240

Sigma-Aldrich

Magnesium chloride hexahydrate

AR, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
AR
form:
crystalline powder
Pricing and availability is not currently available.

grade

AR

product line

Vetec

Assay

≥99%

form

crystalline powder

pH

4.5-7 (5% in water)

anion traces

bromide (Br-): ≤0.05%

cation traces

Ca: ≤0.01%
Fe: ≤0.0002%
heavy metals (as Pb): ≤0.0002%

SMILES string

O.O.O.O.O.O.Cl[Mg]Cl

InChI

1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

DHRRIBDTHFBPNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sameh Herzi et al.
Food research international (Ottawa, Ont.), 116, 145-156 (2019-02-06)
Food grade Water-in-oil-in-water W/O/W double emulsions with high encapsulation efficiency, controlled texture and triggered leakage are usually sought after in many food applications (e.g. functional foods) and still the subject of active research interest. For that purpose, we have used
Elvira Pantuso et al.
Scientific reports, 10(1), 8902-8902 (2020-06-03)
The crystallization of Anti-CD20, a full-length monoclonal antibody, has been studied in the PEG400/Na2SO4/Water system near Liquid-Liquid Phase Separation (LLPS) conditions by both sitting-drop vapour diffusion and batch methods. In order to understand the Anti-CD20 crystallization propensity in the solvent
Chase T Gerold et al.
Analytical chemistry, 90(7), 4894-4900 (2018-03-20)
In this study, paper-based microfluidic devices (μPADs) capable of K+ quantification in aqueous samples, as well as in human serum, using both colorimetric and distance-based methods are described. A lipophilic phase containing potassium ionophore I (valinomycin) was utilized to achieve
Mariusz Sandomierski et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 191, 27-35 (2017-10-06)
Ti6Al4V alloy is the most commonly used in orthopedic industry as an endoprosthesis. Ti6Al4V exhibits good mechanical properties, except the abrasion resistance. Surface modification of Ti6Al4V in order to obtain organic layer, and then the attachment of the polymer, can
Miranda N Emaus et al.
Analytica chimica acta, 1124, 184-193 (2020-06-15)
Multiplex amplification of DNA can be highly valuable in circulating tumor DNA (ctDNA) analysis due to the sheer number of potential mutations. However, commercial ctDNA extraction methods struggle to preconcentrate low concentrations of DNA and require multiple sample handling steps.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service