Skip to Content
Merck
All Photos(1)

Key Documents

V800177

Sigma-Aldrich

Ethyl acetate

LR, ≥99%

Synonym(s):

EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
LR
bp:
76.5-77.5 °C (lit.)
vapor pressure:
73 mmHg ( 20 °C)

grade

LR

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

product line

Vetec

Assay

≥99%

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

dilution

(for analytical testing)

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

solubility

water: soluble

density

0.902 g/mL at 25 °C (lit.)

format

neat

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junwei Liu et al.
Planta, 241(6), 1435-1451 (2015-02-27)
Strigolactone changes and cross talk with ABA unveil a picture of root-specific hormonal dynamics under stress. Strigolactones (SLs) are carotenoid-derived hormones influencing diverse aspects of development and communication with (micro)organisms, and proposed as mediators of environmental stimuli in resource allocation
Yuusuke Fujimoto et al.
PloS one, 7(11), e48685-e48685 (2012-11-13)
Combination therapy with ribavirin, interferon, and viral protease inhibitors could be expected to elicit a high level of sustained virologic response in patients infected with hepatitis C virus (HCV). However, several severe side effects of this combination therapy have been
Tsukasa Horiyama et al.
PloS one, 9(9), e108642-e108642 (2014-09-27)
Escherichia coli produces the iron-chelating compound enterobactin to enable growth under iron-limiting conditions. After biosynthesis, enterobactin is released from the cell. However, the enterobactin export system is not fully understood. Previous studies have suggested that the outer membrane channel TolC
Maarit Neuvonen et al.
PloS one, 9(8), e103743-e103743 (2014-08-27)
Bacterial cholesterol oxidase is commonly used as an experimental tool to reduce cellular cholesterol content. That the treatment also generates the poorly degradable metabolite 4-cholesten-3-one (cholestenone) has received less attention. Here, we investigated the membrane partitioning of cholestenone using simulations
Angela Schwarm et al.
Archives of animal nutrition, 69(3), 159-176 (2015-05-13)
The aim of the present experiment was to compare silage prepared from maize having a brown midrib (BMR) mutation with control (CTR) maize to identify their effects on enteric methane emission, digesta mean retention time (MRT), ruminal fermentation and digestibility.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service