Skip to Content
Merck
All Photos(2)

Key Documents

V800119

Sigma-Aldrich

Chromium(VI) oxide

LR, ≥99%

Synonym(s):

Chromic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CrO3
CAS Number:
Molecular Weight:
99.99
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
Assay:
≥99%
grade:
LR
form:
solid

grade

LR

product line

Vetec

Assay

≥99%

form

solid

pH

<1 (20 °C, 100 g/L)

mp

196 °C (dec.) (lit.)

SMILES string

O=[Cr](=O)=O

InChI

1S/Cr.3O

InChI key

WGLPBDUCMAPZCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 1B - Ox. Sol. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Vayssieres et al.
Journal of nanoscience and nanotechnology, 1(4), 385-388 (2003-08-14)
We are reporting here on the inexpensive fabrication and optical properties of an iron(III) oxide-chromium(III) oxide nanocomposite thin film of corundum crystal structure. Its novel and unique-designed architecture consists of uniformed, well-defined and oriented nanorods of Hematite (alpha-Fe2O3) of 50
S Kondo et al.
European journal of biochemistry, 200(3), 689-698 (1991-09-15)
The chemical structure of the saccharide portion of Vibrio parahaemolyticus serotype 012 lipopolysaccharide was studied. Using chemical degradation and modification, as well as methylation analysis in combination with GLC-MS, laser-desorption mass spectrometry and 1H-NMR and 13C-NMR spectroscopy, the carbohydrate backbone
Seongmin Lee et al.
Organic letters, 11(1), 5-8 (2008-12-06)
The C14,15-dihydro-C22,25-epi north unit of cephalostatin 1 has been synthesized in 11 operations from commercially available hecogenin acetate via multiple reductions and oxidations. The key transformations include (i) Cr(VI)-catalyzed E-ring opening, (ii) C17 hydroxylation, and (iii) a base-triggered cyclization cascade.
Adam Szelag et al.
Polish journal of pharmacology, 55(6), 1097-1103 (2004-01-20)
Hexavalent chromium compounds exhibit higher toxicity than its trivalent compounds since chromium ions in the +6 oxidation state easily cross biological membranes. It has recently been proposed that substances reducing chromium ions from the +6 to the less toxic +3
S Y Shiau et al.
The Journal of nutrition, 123(10), 1747-1753 (1993-10-01)
An experiment was conducted to study the influence of three different forms of chromium (CrCl3.6H2O, Na2CrO4.4H2O and Cr2O3) on the utilization of two carbohydrates (glucose and cornstarch) by juvenile hybrid tilapia (Oreochromis niloticus x O. aureus). Average initial body weight

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service