Skip to Content
Merck
All Photos(1)

Key Documents

V800090

Sigma-Aldrich

Bromothymol Blue

Powder

Synonym(s):

3′,3″-Dibromothymolsulfonphthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H28Br2O5S
CAS Number:
Molecular Weight:
624.38
Beilstein:
373934
EC Number:
MDL number:
UNSPSC Code:
12171502
PubChem Substance ID:
Pricing and availability is not currently available.

Product Name

Bromothymol Blue,

product line

Vetec

form

powder

visual transition interval

5.8-7.6, yellow to blue

mp

200-202 °C (lit.)

λmax

430-435 nm (pH 5.8)
615-618 nm (pH 7.6)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC(C)c1cc(c(C)c(Br)c1O)C2(OS(=O)(=O)c3ccccc23)c4cc(C(C)C)c(O)c(Br)c4C

InChI

1S/C27H28Br2O5S/c1-13(2)17-11-20(15(5)23(28)25(17)30)27(19-9-7-8-10-22(19)35(32,33)34-27)21-12-18(14(3)4)26(31)24(29)16(21)6/h7-14,30-31H,1-6H3

InChI key

NUHCTOLBWMJMLX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hisataka Maruyama et al.
Lab on a chip, 8(2), 346-351 (2008-01-31)
This paper demonstrates local pH measurement in a microchip using a pH-sensing gel-microbead. To achieve this, the gel-microbead made of a hydrophilic photo-crosslinkable resin was functionalized with the pH indicator bromothymol blue (BTB). The primary constituent of this photo-crosslinkable resin
Olga A Zaporozhets et al.
Analytica chimica acta, 597(1), 171-177 (2007-07-31)
Solid-phase spectrophotometric and visual test-methods of fluoride and oxalate determination are proposed. The methods are based on the competitive reactions of ZrOCl2 with methylthymol blue immobilized on silica gel and fluoride or oxalate in solution. Absorbance of the solid-phase reagent
Mariana A Sanchez et al.
Analytica chimica acta, 694(1-2), 95-99 (2011-05-14)
Liquid-liquid microextraction without phase segmentation was implemented in a multicommuted flow system for determination of the anti-hypertensive diltiazem. The procedure was based on ion pair formation between the drug and the dye bromothymol blue at pH 3.5. The detection was
Kristina Jurcic et al.
Journal of chromatography. A, 1134(1-2), 317-325 (2006-10-07)
An effective sample preconcentration technique for proteins and peptides was recently developed using capillary electrophoresis (CE) with discontinuous buffers [C.A. Nesbitt, J.T.-M. Lo, K.K.-C. Yeung, J. Chromatogr. A 1073 (2005) 175]. Two buffers of different pH created a junction to
Bambang Kuswandi et al.
Talanta, 74(4), 613-618 (2008-03-29)
An optical fiber biosensor consisting of acetylcholinesterase (AChE) and bromothymol blue (BTB) doped sol-gel film was employed to detect organophosphate pesticide chlorpyrifos. The main advantage of this optical biosensor is the use of a single sol-gel film with immobilized AChE

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service