Skip to Content
Merck
All Photos(1)

Key Documents

V0T0379

Sigma-Aldrich

Ammonium sulfate

technical grade, 99%

Synonym(s):

Mascagnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

grade

technical grade

Assay

99%

mp

>280 °C (dec.) (lit.)

density

1.77 g/mL at 25 °C (lit.)

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazunori Sugiura et al.
Biochemical and biophysical research communications, 457(3), 242-248 (2015-01-17)
Intracellular redox state is a critical factor for fundamental cellular functions, including regulation of the activities of various metabolic enzymes as well as ROS production and elimination. Genetically-encoded fluorescent redox sensors, such as roGFP (Hanson, G. T., et al. (2004))
James F Hunter et al.
Environmental science & technology, 48(17), 10227-10234 (2014-08-06)
A large number of organic species emitted into the atmosphere contain cycloalkyl groups. While cyclic species are believed to be important secondary organic aerosol (SOA) precursors, the specific role of cyclic moieties (particularly for species with multiple or fused rings)
Yohei Y Yamamoto et al.
Journal of molecular biology, 426(14), 2667-2678 (2014-05-27)
Chaperonins are ubiquitous molecular chaperones with the subunit molecular mass of 60kDa. They exist as double-ring oligomers with central cavities. An ATP-dependent conformational change of the cavity induces the folding of an unfolded protein that is captured in the cavity.
Christian Molitor et al.
Planta, 242(3), 519-537 (2015-02-24)
Aurone synthase belongs to the novel group 2 polyphenol oxidases and the presented kinetic characterization suggests a differing aurone biosynthesis in Asteraceae species compared to snapdragon. Aurone synthases (AUS) are polyphenol oxidases (PPO) physiologically involved in the formation of yellow
Takayoshi Hiyama et al.
The FEBS journal, 281(14), 3113-3125 (2014-05-20)
The last two steps of l-tryptophan (Trp) biosynthesis are catalyzed by Trp synthase, a heterotetramer composed of TrpA and TrpB. TrpB catalyzes the condensation of indole, synthesized by TrpA, and serine to Trp. In the hyperthermophilic archaeon Thermococcus kodakarensis, trpA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service