Skip to Content
Merck
All Photos(1)

Documents

1585006

USP

Pyrazinamide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Pyrazinecarboxamide, Pyrazinamide, Pyrazinoic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N3O
CAS Number:
Molecular Weight:
123.11
Beilstein:
112306
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

pyrazinamide

manufacturer/tradename

USP

mp

189-191 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

NC(=O)c1cnccn1

InChI

1S/C5H5N3O/c6-5(9)4-3-7-1-2-8-4/h1-3H,(H2,6,9)

InChI key

IPEHBUMCGVEMRF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pyrazinamide USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Pyrazinamide Tablets
  • Pyrazinamide Compounded Oral Suspension
  • Rifampin, Isoniazid, and Pyrazinamide Tablets
  • Rifampin, Isoniazid, Pyrazinamide, and Ethambutol Hydrochloride Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service