Skip to Content
Merck
All Photos(1)

Key Documents

1457469

USP

Naratriptan hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Amerge, GR-85548A, N-Methyl-3-(1-methyl-4-piperidinyl)-1H-indole-5-ethanesulfonamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

125 MG
€510.00

€510.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
125 MG
€510.00

About This Item

Empirical Formula (Hill Notation):
C17H25N3O2S ·HCl
CAS Number:
Molecular Weight:
371.93
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€510.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

naratriptan

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CNS(=O)(=O)CCc1ccc2[nH]cc(C3CCN(C)CC3)c2c1

InChI

1S/C17H25N3O2S.ClH/c1-18-23(21,22)10-7-13-3-4-17-15(11-13)16(12-19-17)14-5-8-20(2)9-6-14;/h3-4,11-12,14,18-19H,5-10H2,1-2H3;1H

InChI key

AWEZYKMQFAUBTD-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Naratriptan hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Naratriptan Compounded Oral Suspension
  • Naratriptan Hydrochloride
  • Naratriptan Tablets

Biochem/physiol Actions

Naratriptan hydrochloride is a serotonin 5-HT1B/1D receptor agonist.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service