Skip to Content
Merck
All Photos(1)

Documents

1437600

USP

Methylsulfonylmethane

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Dimethyl sulfone, Methyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO2
CAS Number:
Molecular Weight:
94.13
Beilstein:
1737717
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

methylsulfonylmethane

manufacturer/tradename

USP

bp

238 °C (lit.)

mp

107-109 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CS(C)(=O)=O

InChI

1S/C2H6O2S/c1-5(2,3)4/h1-2H3

InChI key

HHVIBTZHLRERCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methylsulfonylmethane USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youn Hee Joung et al.
International journal of oncology, 44(3), 883-895 (2014-01-10)
Human urinary bladder cancer is the fifth most common cancer, with a worldwide estimate of about two million patients. Recurrence after complete transurethral prostatic resection is the most important problem in therapy. Combination therapy is a new approach in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service