Skip to Content
Merck
All Photos(1)

Key Documents

1255850

USP

Eszopiclone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(+)-Eszopiclone, (+)-(5S)-6-(5-Chloropyridin-2-yl)-7-oxo-6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5-yl 4-methylpiperazine-1-carboxylate, (S)-Zopiclone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17ClN6O3
CAS Number:
Molecular Weight:
388.81
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Product 1255850 is not currently sold in your country. Contact Technical Service

grade

pharmaceutical primary standard

API family

eszopiclone

manufacturer/tradename

USP

drug control

USDEA Schedule IV

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Clc1cnc(cc1)N2[C@H](c4nccnc4C2=O)OC(=O)N3CCN(CC3)C

InChI

1S/C17H17ClN6O3/c1-22-6-8-23(9-7-22)17(26)27-16-14-13(19-4-5-20-14)15(25)24(16)12-3-2-11(18)10-21-12/h2-5,10,16H,6-9H2,1H3/t16-/m0/s1

InChI key

GBBSUAFBMRNDJC-INIZCTEOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Eszopiclone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Eszopiclone Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hi. What is the solubility of Eszopiclone in water and PBS?

    1 answer
    1. The solubility of this USP standard has not been determined. Various sources note that this material is soluble at 0.885 mg/mL in water. In addition, PBS at pH 3.2 and 0.01 M HCl are listed, with no concentration reported. Please see the links below to review additional information that may be helpful:
      https://pubs.acs.org/doi/10.1021/acs.jced.0c00047#
      https://go.drugbank.com/drugs/DB00402
      https://patents.google.com/patent/EP1997482A1/en

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service