1225678
USP
Doxofylline Related Compound B
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
1-[(1,3-Dioxolan-2-yl)methyl]-N-methyl-4-(methylamino)-1H-imidazole-5-carboxamide
About This Item
Recommended Products
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
storage temp.
2-8°C
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service