Skip to Content
Merck
All Photos(1)

Key Documents

1154707

USP

Cyclomethicone 4

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Octamethylcyclotetrasiloxane

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
€431.00

€431.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
200 MG
€431.00

About This Item

Linear Formula:
[-Si(CH3)2O-]4
CAS Number:
Molecular Weight:
296.62
Beilstein:
1787074
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€431.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

vapor density

1 (vs air)

API family

cyclomethicone

form

liquid

manufacturer/tradename

USP

refractive index

n20/D 1.396 (lit.)

bp

175-176 °C (lit.)

mp

17-18 °C (lit.)

density

0.956 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1

InChI

1S/C8H24O4Si4/c1-13(2)9-14(3,4)11-16(7,8)12-15(5,6)10-13/h1-8H3

InChI key

HMMGMWAXVFQUOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service