Skip to Content
Merck
All Photos(1)

Key Documents

1118361

USP

Chloroquine Related Compound E

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N4-(5-Chloroquinolin-4-yl)-N1,N1-diethylpentane-1,4-diamine oxalate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€1,380.00

€1,380.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€1,380.00

About This Item

Empirical Formula (Hill Notation):
C20H28ClN3O4
CAS Number:
Molecular Weight:
409.91
UNSPSC Code:
41116107
NACRES:
NA.24

€1,380.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

synthetic

grade

pharmaceutical primary standard

Agency

USP

API family

chloroquine

form

solid

packaging

pkg of 10 mg

manufacturer/tradename

USP

storage condition

protect from light

color

white

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Clc1c2c(nccc2NC(CCCN(CC)CC)C)ccc1

InChI

1S/C18H26ClN3/c1-4-22(5-2)13-7-8-14(3)21-17-11-12-20-16-10-6-9-15(19)18(16)17/h6,9-12,14H,4-5,7-8,13H2,1-3H3,(H,20,21)

InChI key

CHDBGFBFAXJWRC-UHFFFAOYSA-N

General description

Chloroquine Related Compound E is an impurity of chloroquine, which is a 9-aminoquinoline drug that effectively inhibits viral infections in humans.[1]
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chloroquine Related Compound E can be used as a pharmaceutical primary standard for the determination of the analyte in pharmaceutical formulations by liquid chromatography (LC) technique
Also, for use with USP monograph such as Chloroquine Phosphate.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chloroquine Phosphate
Pharmacopeia, US
United States Pharmacopeia, 39(3), 959-959 (2020)
Chloroquine is a potent inhibitor of SARS coronavirus infection and spread
Vincent MJ, et al.
Virology Journal, 2(1), 1-10 (2005)

Protocols

Chloroquine Phosphate HPLC Assay meets system suitability criteria with good resolution, reproducibility, and sensitivity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service