Skip to Content
Merck
All Photos(1)

Key Documents

1071348

USP

Bimatoprost

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

((Z)-7-[(1R,2R,3R,5S)-3,5-Dihydroxy-2-[(S,E)-3-hydroxy-5-phenylpent-1-en-1-yl]cyclopentyl]-Nethylhept-5-enamide)

Sign Into View Organizational & Contract Pricing

Select a Size

1071348-100MG
€2,530.00

€2,530.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1071348-100MG
€2,530.00

About This Item

Empirical Formula (Hill Notation):
C25H37NO4
CAS Number:
Molecular Weight:
415.57
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.24

€2,530.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

Agency

USP

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

storage temp.

−20°C

SMILES string

N(CC)C(=O)CCC\C=C/C[C@@H]1[C@H]([C@@H](C[C@@H]1O)O)\C=C\[C@@H](O)CCc2ccccc2

InChI key

AQOKCDNYWBIDND-FTOWTWDKSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

USP issued SDS can be found here
Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service