Skip to Content
Merck
All Photos(1)

Documents

SY3H998DB754

Synthonix Corporation

2-methyl-3-oxo-2,4-dihydro-1H-quinoxaline-6-carbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
190.2
MDL number:

Iupac Name

2-methyl-3-oxo-1,2,3,4-tetrahydroquinoxaline-6-carbaldehyde

Inchi Code

1S/C10H10N2O2/c1-6-10(14)12-9-4-7(5-13)2-3-8(9)11-6/h2-6,11H,1H3,(H,12,14)

InChI key

GMMGMRUFNMLCOX-UHFFFAOYSA-N

storage temp.

Refrigerator

Purity

98%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

solid

Product Link

https://synthonix.com/80170.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://synthonix.com/80170.html


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service