Skip to Content
Merck
All Photos(1)

Key Documents

SY3H11AB2000

Synthonix Corporation

4-Iodotetrahydrothiopyran

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
228.1
MDL number:

Iupac Name

4-iodotetrahydro-2H-thiopyran

Inchi Code

1S/C5H9IS/c6-5-1-3-7-4-2-5/h5H,1-4H2

InChI key

JVBIONNYIWEILC-UHFFFAOYSA-N

storage temp.

Refrigerator

Purity

97%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Liquid

Product Link

https://synthonix.com/31818.html

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library