Skip to Content
Merck
All Photos(1)

Key Documents

Z777221

Proteomix® SAX-NP3 (3 µm) HPLC Columns

L × I.D. 5 cm × 2.1 mm, PEEK

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€1,500.00

€1,500.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€1,500.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

€1,500.00


Please contact Customer Service for Availability

Product Name

Proteomix® SAX-NP3 Strong Anion Exchange Column, L × I.D. 5 cm × 2.1 mm, 3 μm particle size, PEEK

material

PEEK

Agency

suitable for USP L50

product line

Proteomix®

manufacturer/tradename

Sepax 403NP3P-2105

packaging

1 ea of

parameter

55 Mpa max. pressure (8000 psi)
80 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

5 cm × 2.1 mm

matrix

spherical PS-DVB
polymeric

matrix active group

ammonium ion, quaternary phase

particle size

3 μm

pore size

non-porous

operating pH range

2.0-12.0

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Proteomix columns are packed with poly(styrene divinylbenzene) particles that are shielded by a hydrophilic polymer layer that contains a network of weak (WAX) or strong (SAX) anion exchange groups. Use Proteomix anion-exchange columns for the analyis or small scale purification of negatively charged biopolymers, such as glycoprotein isoforms, pegylated proteins and oligonucleotides.

Proteomix User Manual

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Proteomix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service