Skip to Content
Merck
All Photos(1)

Documents

Z777075

Zenix®-C SEC-100 (3 µm) HPLC Columns

L x I.D. 30 cm × 7.8 mm, hydrophilic polymer phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

product name

Zenix®-C SEC-100 Gel Filtration Column, 3 μm, L × I.D. 30 cm × 7.8 mm, 3 μm particle size, stainless steel

material

stainless steel

Agency

suitable for USP L59

product line

Zenix®

mol wt

100-100,000

manufacturer/tradename

Sepax 233100-7830

packaging

1 ea of

parameter

20 Mpa max. pressure
80 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

30 cm × 7.8 mm

impurities

<10 ppm metals

matrix

spherical silica
fully porous particle

matrix active group

hydrophilic polymer phase

particle size

3 μm

pore size

100 Å

operating pH range

2.0-8.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The bonded phase in Zenix-C SEC columns is more polar than the hydrophilic phase used in Zenix columns, making Zenix-C columns better suited for the gel filtration analysis of more hydrophobic bioopolymers such as membrane proteins and pegylated antibodies. Both Zenix-C and Zenix columns are based on the same high efficiency 3 micron particles and each product line is available in the same wide range of pore sizes to tailor the optimal pore size to the hydrodynamic radius of the biopolymer of interest.

Zenix User Manual

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Zenix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service