product line
univAR™
packaging
poly bottle of 12 × 500 g (SAAR5046020EM)
mp
334 °C (lit.)
storage temp.
2-30°C
SMILES string
[K+].[O-][N+]([O-])=O
InChI
1S/K.NO3/c;2-1(3)4/q+1;-1
InChI key
FGIUAXJPYTZDNR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Analysed reagent
For laboratory use only
Legal Information
univAR is a trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Ox. Sol. 3
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service