Skip to Content
Merck
All Photos(1)

Key Documents

SAAR4946130

Perchloric acid solution

0.1 N, 0.1 M

Synonym(s):

Perchloric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
MDL number:
UNSPSC Code:
12352301

packaging

poly bottle of 1 × 500 mL (SAAR4946130KF)

concentration

0.1 M
0.1 N

storage temp.

2-30°C

SMILES string

OCl(=O)(=O)=O

InChI

1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)

InChI key

VLTRZXGMWDSKGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For laboratory use only

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.0 °F

Flash Point(C)

40 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service